The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288625.1 | NP_001275554.1 | 3 - 4 | 1024 | 105 |
NM_001288626.1 | NP_001275555.1 | 3 - 4 | 1178 | 105 | |
NM_007880.4 | NP_031906.1 | 3 - 4 | 1178 | 105 | |
GenBank mRNA | AK141283.1 | - | 3 - 4 | 1025 | 105 |
AK154956.1 | - | 3 - 4 | 1150 | 105 | |
BC050925.1 | - | 3 - 4 | 1082 | 105 | |
U60335.1 | - | 2 - 3 | 966 | 105 |