The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007881.4 | NP_031907.2 | 4 - 5 | 617 | 71 |
GenBank mRNA | BC050920.1 | - | 4 - 5 | 617 | 71 |
BC053051.1 | - | 4 - 5 | 445 | 71 | |
D87744.1 | - | 3 - 4 | 327 | 71 |