The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008299.3 | NP_032325.2 | 1 - 1 | 651 | 80 |
GenBank mRNA | AK005688.1 | - | 1 - 1 | 609 | 80 |
AK005690.1 | - | 1 - 1 | 600 | 80 | |
BC048490.1 | - | 1 - 1 | 619 | 80 | |
U95607.1 | - | 1 - 1 | 653 | 80 |