The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008405.3 | NP_032431.2 | 12 - 13 | 1656 | 98 |
GenBank mRNA | AF051367.1 | - | 12 - 13 | 1641 | 98 |
AK036640.1 | - | 12 - 13 | 1656 | 98 | |
AK137534.1 | - | 12 - 13 | 1657 | 98 | |
BC113142.1 | - | 12 - 13 | 1625 | 98 |