The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008444.4 | NP_032470.3 | 2 - 3 | 1554 | 65 |
GenBank mRNA | AK083700.1 | - | 2 - 3 | 1542 | 65 |
AK122258.1 | - | 2 - 3 | 1515 | 65 | |
AK147467.1 | - | 2 - 3 | 1548 | 65 | |
AK148200.1 | - | 2 - 3 | 1535 | 65 | |
BC139388.1 | - | 1 - 2 | 1473 | 65 | |
BC139389.1 | - | 1 - 2 | 1473 | 65 | |
D26077.1 | - | 1 - 2 | 1467 | 65 |