The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008817.2 | NP_032843.2 | 9 - 9 | 3098 | 67 |
GenBank mRNA | AB003040.1 | - | 9 - 9 | 3098 | 67 |
AF038939.1 | - | 6 - 6 | 2749 | 67 | |
AK044602.1 | - | 10 - 10 | 3162 | 67 | |
AK122238.1 | - | 9 - 9 | 3077 | 67 | |
AK141217.1 | - | 9 - 9 | 3099 | 67 | |
AK163845.1 | - | 9 - 9 | 3101 | 67 | |
BC048778.1 | - | 4 - 4 | 2475 | 67 | |
BC072661.1 | - | 9 - 9 | 3086 | 67 | |
BC085183.1 | - | 9 - 9 | 3083 | 67 | |
BC089344.1 | - | 1 - 1 | 2178 | 67 |