The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009319.3 | NP_033345.2 | 1 - 2 | 413 | 60 |
GenBank mRNA | AK143188.1 | - | 1 - 2 | 414 | 60 |
AV454829.1 | - | 1 - 2 | 101 | 60 | |
AV582078.1 | - | 1 - 2 | 271 | 60 | |
BC002028.1 | - | 1 - 2 | 126 | 60 | |
BI737851.1 | - | 1 - 2 | 360 | 60 | |
BQ948575.1 | - | 1 - 2 | 154 | 60 | |
U79962.1 | - | 1 - 2 | 146 | 60 |