Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc3
Entrez Gene ID
22129
Gene Name
tetratricopeptide repeat domain 3
Gene Aliases
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119
Chromosome Location
Chr.16: 94370739 - 94469222 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009441.2 NP_033467.2 28 - 29 3127 68
GenBank mRNA AB008516.1 - 27 - 28 3004 68
AK088273.1 - 27 - 28 3074 68
AK148494.1 - 1 - 2 151 68
AK163496.1 - 2 - 3 432 68
AK165850.1 - 17 - 18 2035 68
AK166120.1 - 18 - 19 2067 68
AK220495.1 - 28 - 29 3169 68
BC019173.1 - 28 - 29 3096 68
BC037654.1 - 27 - 28 3027 68
BC057207.1 - 27 - 28 3031 68
BC145489.1 - 23 - 24 2647 68

View Full Product Details