Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xrcc1
Entrez Gene ID
22594
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 1
Gene Aliases
Xrcc-1
Chromosome Location
Chr.7: 24546300 - 24573438 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009532.4 NP_033558.3 7 - 8 877 61
GenBank mRNA AK155376.1 - 7 - 8 854 61
AK165569.1 - 7 - 8 832 61
AK168334.1 - 7 - 8 827 61
AK168727.1 - 7 - 8 831 61
AK217480.1 - 1 - 2 120 61
BC055900.1 - 7 - 8 853 61
BC085281.1 - 9 - 10 1259 61
U02887.1 - 7 - 8 841 61

View Full Product Details