Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xrcc1
Entrez Gene ID
22594
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 1
Gene Aliases
Xrcc-1
Chromosome Location
Chr.7: 24546300 - 24573438 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009532.4 NP_033558.3 10 - 11 1355 85
GenBank mRNA AK155376.1 - 10 - 11 1332 85
AK165569.1 - 10 - 11 1310 85
AK168334.1 - 10 - 11 1305 85
AK168727.1 - 10 - 11 85
BC055900.1 - 10 - 11 1331 85
BC085281.1 - 12 - 13 1737 85
U02887.1 - 10 - 11 1319 85

View Full Product Details