The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289612.1 | NP_001276541.1 | 4 - 5 | 870 | 89 |
NM_001289765.1 | NP_001276694.1 | 11 - 12 | 1817 | 89 | |
NM_001289766.1 | NP_001276695.1 | 11 - 12 | 1782 | 89 | |
NM_009539.3 | NP_033565.2 | 11 - 12 | 1774 | 89 | |
GenBank mRNA | AB083210.1 | - | 4 - 5 | 873 | 89 |
AK039883.1 | - | 11 - 12 | 1711 | 89 | |
AK089056.1 | - | 11 - 12 | 1746 | 89 | |
BC029727.1 | - | 11 - 12 | 1732 | 89 | |
U04379.1 | - | 11 - 12 | 89 | ||
U77667.1 | - | 10 - 11 | 1620 | 89 |