The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191023.1 | NP_001177952.1 | 11 - 12 | 1531 | 73 |
NM_001191025.1 | NP_001177954.1 | 11 - 12 | 1480 | 73 | |
NM_001191026.1 | NP_001177955.1 | 12 - 13 | 1507 | 73 | |
NM_001191027.1 | NP_001177956.1 | 11 - 12 | 1447 | 73 | |
NM_010063.4 | NP_034193.2 | 12 - 13 | 1540 | 73 | |
GenBank mRNA | AF063229.1 | - | 12 - 13 | 1374 | 73 |
AF063230.1 | - | 11 - 12 | 1314 | 73 | |
AK158589.1 | - | 12 - 13 | 1508 | 73 | |
BC051992.1 | - | 12 - 13 | 1268 | 73 | |
GU992206.1 | - | 11 - 12 | 1272 | 73 | |
GU992207.1 | - | 11 - 12 | 1188 | 73 |