Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gem
Entrez Gene ID
14579
Gene Name
GTP binding protein (gene overexpressed in skeletal muscle)
Gene Aliases
AV020497
Chromosome Location
Chr.4: 11704447 - 11714993 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010276.4 NP_034406.2 4 - 5 746 77
GenBank mRNA AK157655.1 - 4 - 5 748 77
AK158393.1 - 4 - 5 748 77
AK168401.1 - 4 - 5 747 77
AK171336.1 - 4 - 5 684 77
BC029668.1 - 4 - 5 722 77
U10551.1 - 3 - 4 725 77
U13053.1 - 4 - 5 737 77

View Full Product Details