The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010296.2 | NP_034426.2 | 2 - 3 | 139 | 68 |
GenBank mRNA | AB025922.1 | - | 2 - 3 | 149 | 68 |
AB234617.1 | - | 2 - 3 | 531 | 68 | |
AF026305.1 | - | 2 - 3 | 139 | 68 | |
BC131649.1 | - | 1 - 2 | 117 | 68 | |
BC131650.1 | - | 1 - 2 | 117 | 68 | |
CJ062804.1 | - | 1 - 2 | 123 | 68 |