The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010324.2 | NP_034454.2 | 7 - 8 | 1108 | 60 |
GenBank mRNA | AK146445.1 | - | 7 - 8 | 1010 | 60 |
AK147807.1 | - | 7 - 8 | 1031 | 60 | |
AK147934.1 | - | 7 - 8 | 1013 | 60 | |
BC002057.1 | - | 7 - 8 | 996 | 60 | |
J02623.1 | - | 7 - 8 | 1012 | 60 |