The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010657.3 | NP_034787.2 | 8 - 9 | 7193 | 68 |
GenBank mRNA | AK173200.1 | - | 5 - 6 | 2439 | 68 |
AY454345.1 | - | 9 - 10 | 7140 | 68 | |
BC145272.1 | - | 4 - 5 | 6536 | 68 | |
L07911.1 | - | 5 - 6 | 1794 | 68 |