The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011543.4 | NP_035673.3 | 2 - 3 | 220 | 118 |
GenBank mRNA | AF083214.1 | - | 2 - 3 | 144 | 118 |
AK002983.1 | - | 2 - 3 | 191 | 118 | |
AK010628.1 | - | 2 - 3 | 206 | 118 | |
AK012498.1 | - | 2 - 3 | 179 | 118 | |
AK014245.1 | - | 2 - 3 | 179 | 118 | |
AK027909.1 | - | 2 - 3 | 178 | 118 | |
AK078330.1 | - | 2 - 3 | 206 | 118 | |
AK088339.1 | - | 2 - 3 | 179 | 118 | |
AK152609.1 | - | 2 - 3 | 178 | 118 | |
AK160198.1 | - | 2 - 3 | 179 | 118 | |
AK166673.1 | - | 2 - 3 | 179 | 118 | |
BC002115.1 | - | 2 - 3 | 163 | 118 |