Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dsc1
Entrez Gene ID
13505
Gene Name
desmocollin 1
Gene Aliases
1110020A10Rik, AI507491, Dsc1a, Dsc1b
Chromosome Location
Chr.18: 20083471 - 20114773 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291804.1 NP_001278733.1 2 - 3 368 70
NM_013504.5 NP_038532.3 2 - 3 368 70
GenBank mRNA AK036893.1 - 2 - 3 254 70
AK079441.1 - 2 - 3 369 70
AK081826.1 - 2 - 3 276 70
BC109161.1 - 2 - 3 205 70
BC109162.1 - 2 - 3 205 70
X97986.1 - 2 - 3 302 70

View Full Product Details