The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016873.2 | NP_058569.2 | 1 - 2 | 273 | 62 |
GenBank mRNA | AF100778.1 | - | 1 - 2 | 220 | 62 |
AF126063.1 | - | 1 - 2 | 205 | 62 | |
BC032877.1 | - | 1 - 2 | 198 | 62 | |
BQ560868.1 | - | 1 - 2 | 273 | 62 |