Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ybx2
Entrez Gene ID
53422
Gene Name
Y box protein 2
Gene Aliases
Msy2
Chromosome Location
Chr.11: 69935835 - 69941604 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016875.2 NP_058571.2 8 - 9 1189 86
GenBank mRNA AF073954.1 - 8 - 9 1116 86
AF073955.1 - 8 - 9 983 86
AK131732.1 - 2 - 3 308 86
AK161439.1 - 4 - 5 581 86
BC141112.1 - 8 - 9 1292 86

View Full Product Details