The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016978.2 | NP_058674.1 | 4 - 5 | 665 | 81 |
GenBank mRNA | AK128914.1 | - | 4 - 5 | 656 | 81 |
AK145877.1 | - | 4 - 5 | 666 | 81 | |
AK146413.1 | - | 4 - 5 | 610 | 81 | |
AK168851.1 | - | 4 - 5 | 614 | 81 | |
BC008119.1 | - | 4 - 5 | 598 | 81 | |
X64837.1 | - | 3 - 4 | 639 | 81 |