Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmeb1
Entrez Gene ID
56809
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
1110050A04Rik, AI256615, AI481278
Chromosome Location
Chr.4: 132220376 - 132261549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122992.1 NP_001116464.1 9 - 10 1016 81
NM_020273.2 NP_064669.2 8 - 9 953 81
GenBank mRNA AF210433.1 - 7 - 8 914 81
AK134349.1 - 8 - 9 954 81
BC046983.1 - 9 - 10 987 81

View Full Product Details