Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankra2
Entrez Gene ID
68558
Gene Name
ankyrin repeat, family A (RFXANK-like), 2
Gene Aliases
1110004M18Rik, Ankra
Chromosome Location
Chr.13: 98263076 - 98274754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271388.1 NP_001258317.1 7 - 8 1295 105
NM_001271389.1 NP_001258318.1 7 - 8 1295 105
NM_001271390.1 NP_001258319.1 6 - 7 913 105
NM_001271391.1 NP_001258320.1 6 - 7 913 105
NM_001271392.1 NP_001258321.1 6 - 7 913 105
NM_023472.2 NP_075961.1 7 - 8 1295 105
GenBank mRNA AB221648.1 - 6 - 7 798 105
AF314031.1 - 7 - 8 1129 105
AK003438.2 - 6 - 7 832 105
AK054037.1 - 7 - 8 1101 105
AK081695.1 - 6 - 7 798 105
AK139343.1 - 7 - 8 1177 105
AK161446.1 - 7 - 8 1301 105
BC037437.1 - 6 - 7 841 105
BC066113.1 - 6 - 7 832 105
BC079615.1 - 7 - 8 1112 105
BF730624.1 - 2 - 3 105

View Full Product Details