Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps28
Entrez Gene ID
66914
Gene Name
vacuolar protein sorting 28 (yeast)
Gene Aliases
1110014J03Rik, CIIA, D730005C08Rik
Chromosome Location
Chr.15: 76622088 - 76626027 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF373710.1 - 8 - 9 576 81
AK003572.2 - 8 - 9 606 81
AK003699.1 - 8 - 9 530 81
AK013852.1 - 8 - 9 527 81
BC013535.1 - 8 - 9 539 81

View Full Product Details