The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031873.1 | NP_114079.1 | 5 - 6 | 1631 | 56 |
GenBank mRNA | AF337041.1 | - | 5 - 6 | 1631 | 56 |
BC137740.1 | - | 6 - 7 | 1533 | 56 | |
BC144879.1 | - | 6 - 7 | 1533 | 56 |