Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hps3
Entrez Gene ID
12807
Gene Name
Hermansky-Pudlak syndrome 3 homolog (human)
Gene Aliases
coa
Chromosome Location
Chr.3: 19995945 - 20035376 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146323.1 NP_001139795.1 15 - 16 2437 97
NM_080634.4 NP_542365.3 15 - 16 2833 97
GenBank mRNA AF393747.1 - 15 - 16 2822 97
AF393780.1 - 15 - 16 2825 97
AK034377.1 - 15 - 16 2804 97
AK036663.1 - 15 - 16 2841 97
AK155497.1 - 15 - 16 2801 97
AK157953.1 - 15 - 16 2819 97
AK169855.1 - 14 - 15 2905 97
AK170581.1 - 15 - 16 2349 97
AK171997.1 - 15 - 16 2425 97
BC020105.1 - 5 - 6 932 97
BC038087.1 - 5 - 6 924 97
BC058235.1 - 15 - 16 2807 97

View Full Product Details