Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pask
Entrez Gene ID
269224
Gene Name
PAS domain containing serine/threonine kinase
Gene Aliases
mKIAA0135
Chromosome Location
Chr.1: 93309437 - 93343350 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080850.2 NP_543126.2 8 - 9 1615 79
GenBank mRNA AK028435.1 - 8 - 9 1617 79
AK083528.1 - 8 - 9 1586 79
AK083590.1 - 8 - 9 1580 79
AK129063.1 - 8 - 9 1505 79
AK172031.1 - 8 - 9 1563 79
BC150821.1 - 7 - 8 1327 79

View Full Product Details