The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042534.3 | NP_001035999.1 | 6 - 7 | 913 | 113 |
NM_001271395.1 | NP_001258324.1 | 6 - 7 | 906 | 113 | |
NM_001271415.1 | NP_001258344.1 | 7 - 8 | 903 | 113 | |
NM_007599.4 | NP_031625.2 | 6 - 7 | 760 | 113 | |
GenBank mRNA | AK146622.1 | - | 6 - 7 | 764 | 113 |
AK149857.1 | - | 6 - 7 | 710 | 113 | |
AK152818.1 | - | 6 - 7 | 738 | 113 | |
AK159577.1 | - | 6 - 7 | 713 | 113 | |
AK159736.1 | - | 6 - 7 | 715 | 113 | |
AK165153.1 | - | 6 - 7 | 727 | 113 | |
AK165586.1 | - | 4 - 5 | 556 | 113 | |
BC003480.1 | - | 6 - 7 | 714 | 113 | |
BC023101.1 | - | 6 - 7 | 702 | 113 | |
X54511.1 | - | 7 - 8 | 902 | 113 |