The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008881.2 | NP_032907.1 | 3 - 4 | 1650 | 111 |
GenBank mRNA | BC138023.1 | - | 2 - 3 | 1442 | 111 |
D86948.1 | - | 3 - 4 | 1634 | 111 |