The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135657.1 | NP_001129129.1 | 8 - 9 | 1690 | 75 |
NM_008982.5 | NP_033008.3 | 8 - 9 | 1974 | 75 | |
GenBank mRNA | AK147318.1 | - | 8 - 9 | 1895 | 75 |
AK147556.1 | - | 8 - 9 | 1895 | 75 | |
AY038891.1 | - | 8 - 9 | 1792 | 75 | |
AY039232.1 | - | 8 - 9 | 1792 | 75 | |
D45212.1 | - | 8 - 9 | 1930 | 75 | |
D83203.1 | - | 8 - 9 | 1691 | 75 | |
DQ133576.1 | - | 8 - 9 | 1705 | 75 |