Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myb
Entrez Gene ID
17863
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Chromosome Location
Chr.10: 21124930 - 21160984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198914.1 NP_001185843.1 15 - 16 2420 108
NM_010848.3 NP_034978.3 14 - 15 2063 108
GenBank mRNA AK088020.1 - 1 - 2 93 108
AK088079.1 - 15 - 16 2341 108
AK132250.1 - 14 - 15 1928 108
AK137794.1 - 8 - 9 1008 108
BC011513.1 - 14 - 15 1971 108
M12848.1 - 14 - 15 2061 108
X02774.1 - 14 - 15 1833 108

View Full Product Details