The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040695.1 | NP_001035785.1 | 1 - 2 | 131 | 69 |
GenBank mRNA | AK154536.1 | - | 1 - 2 | 82 | 69 |
AK155685.1 | - | 1 - 2 | 78 | 69 | |
U97571.1 | - | 1 - 2 | 85 | 69 |