The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019803.3 | NP_062777.2 | 3 - 4 | 181 | 102 |
GenBank mRNA | AF296657.1 | - | 3 - 4 | 162 | 102 |
AK136365.1 | - | 3 - 4 | 168 | 102 | |
AK146554.1 | - | 3 - 4 | 186 | 102 | |
AK154464.1 | - | 3 - 4 | 202 | 102 | |
BC010321.1 | - | 3 - 4 | 181 | 102 | |
U93241.1 | - | 2 - 3 | 81 | 102 |