The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167799.1 | NP_001161271.1 | 2 - 3 | 400 | 65 |
NM_019829.4 | NP_062803.4 | 1 - 2 | 699 | 65 | |
GenBank mRNA | AF232709.1 | - | 1 - 2 | 67 | 65 |
AK002722.1 | - | 2 - 3 | 382 | 65 | |
AK166087.1 | - | 1 - 2 | 702 | 65 | |
BC004849.1 | - | 2 - 3 | 333 | 65 | |
BC021883.1 | - | 1 - 2 | 682 | 65 | |
CN539678.1 | - | 2 - 3 | 364 | 65 |