Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Piwil2
Entrez Gene ID
57746
Gene Name
piwi-like RNA-mediated gene silencing 2
Gene Aliases
Piwil1l, mili
Chromosome Location
Chr.14: 70372477 - 70436916 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021308.1 NP_067283.1 16 - 17 2178 96
GenBank mRNA AB032605.1 - 16 - 17 2178 96
AF285586.1 - 7 - 8 907 96
AK030116.1 - 16 - 17 2122 96
AK163647.1 - 3 - 4 433 96
BC138444.1 - 16 - 17 2166 96
BC145717.1 - 16 - 17 2166 96

View Full Product Details