The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023523.5 | NP_076012.3 | 1 - 2 | 221 | 94 |
GenBank mRNA | AF232011.1 | - | 1 - 2 | 127 | 94 |
AF242204.1 | - | 1 - 2 | 161 | 94 | |
AK010260.1 | - | 1 - 2 | 212 | 94 | |
AK090327.1 | - | 1 - 2 | 166 | 94 | |
AK135714.1 | - | 1 - 2 | 166 | 94 | |
AK162100.1 | - | 1 - 2 | 169 | 94 | |
BC013530.1 | - | 1 - 2 | 145 | 94 | |
BG078542.2 | - | 1 - 2 | 221 | 94 |