The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025272.2 | NP_079548.1 | 3 - 4 | 362 | 76 |
GenBank mRNA | AK007610.1 | - | 3 - 4 | 341 | 76 |
AK008464.1 | - | 3 - 4 | 342 | 76 | |
AK013229.1 | - | 3 - 4 | 323 | 76 | |
AK017760.1 | - | 3 - 4 | 364 | 76 | |
AK030335.1 | - | 3 - 4 | 314 | 76 | |
AK159669.1 | - | 3 - 4 | 350 | 76 | |
AK168015.1 | - | 3 - 4 | 331 | 76 | |
BC024733.1 | - | 3 - 4 | 326 | 76 | |
BY707658.1 | - | 3 - 4 | 75 |