The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025313.2 | NP_079589.2 | 1 - 2 | 294 | 64 |
GenBank mRNA | AA409366.1 | - | 1 - 2 | 298 | 64 |
AK002330.1 | - | 1 - 2 | 244 | 64 | |
AK011374.1 | - | 1 - 2 | 215 | 64 | |
AK018011.1 | - | 1 - 2 | 240 | 64 | |
AK076064.1 | - | 1 - 2 | 219 | 64 | |
AK151151.1 | - | 1 - 2 | 229 | 64 | |
AK155285.1 | - | 1 - 2 | 278 | 64 | |
AK165932.1 | - | 1 - 2 | 243 | 64 | |
BC008273.1 | - | 1 - 2 | 203 | 64 | |
CT010177.1 | - | 1 - 2 | 143 | 64 |