The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026703.2 | NP_080979.1 | 1 - 2 | 95 | 72 |
GenBank mRNA | AK002744.1 | - | 1 - 2 | 95 | 72 |
BC012416.1 | - | 1 - 2 | 93 | 72 | |
BC024673.1 | - | 1 - 2 | 92 | 72 |