Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tlcd1
Entrez Gene ID
68385
Gene Name
TLC domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 78176716 - 78180820 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026708.2 NP_080984.1 1 - 2 402 75
GenBank mRNA AK002703.1 - 1 - 2 298 75
AK046577.1 - 2 - 3 2282 75
AK161721.1 - - 302 75
BC005702.1 - 1 - 2 294 75

View Full Product Details