The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026855.4 | NP_081131.1 | 3 - 4 | 481 | 76 |
GenBank mRNA | AK004398.1 | - | 3 - 4 | 482 | 76 |
AK042561.1 | - | 1 - 2 | 83 | 76 | |
BC119376.1 | - | 3 - 4 | 461 | 76 | |
BC120631.1 | - | 3 - 4 | 461 | 76 | |
BC144876.1 | - | 3 - 4 | 421 | 76 |