The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159559.1 | NP_001153031.1 | 3 - 4 | 404 | 61 |
NM_026858.3 | NP_081134.2 | 3 - 4 | 404 | 61 | |
GenBank mRNA | AK004405.1 | - | 3 - 4 | 384 | 61 |
AK010471.1 | - | 3 - 4 | 387 | 61 | |
AK039002.1 | - | 3 - 4 | 385 | 61 | |
BC030630.1 | - | 3 - 4 | 366 | 61 | |
BG801921.1 | - | 3 - 4 | 404 | 61 |