Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Anln
Entrez Gene ID
68743
Gene Name
anillin, actin binding protein
Gene Aliases
1110037A17Rik, 2900037I21Rik, C78101, Scraps
Chromosome Location
Chr.9: 22331214 - 22389696 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028390.3 NP_082666.2 17 - 18 2915 110
GenBank mRNA AK045703.1 - 14 - 15 1879 110
AK046102.1 - 6 - 7 701 110
BC030489.1 - 10 - 11 1257 110
BC032164.1 - 17 - 18 2933 110

View Full Product Details