Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xpo6
Entrez Gene ID
74204
Gene Name
exportin 6
Gene Aliases
2610005L19Rik, AI854665, AL022631, C230091E20Rik, R75304, Ranbp20, mKIAA0370
Chromosome Location
Chr.7: 126101718 - 126200496 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028816.2 NP_083092.2 22 - 23 3540 79
GenBank mRNA AB093235.1 - 17 - 18 3805 79
AK011324.2 - 4 - 5 1896 79
AK154709.1 - 22 - 23 3585 79
AY029528.1 - 21 - 22 3036 79
BC058090.1 - 22 - 23 3540 79

View Full Product Details