The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029095.2 | NP_083371.2 | 9 - 10 | 1300 | 61 |
NR_027967.1 | 9 - 10 | 1180 | 61 | ||
GenBank mRNA | AK003605.1 | - | 9 - 10 | 1177 | 61 |
AK157924.1 | - | 9 - 10 | 1098 | 61 | |
BC024464.1 | - | 9 - 10 | 1156 | 61 |