The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133660.3 | NP_598421.1 | 1 - 2 | 246 | 63 |
GenBank mRNA | AK144549.1 | - | 1 - 2 | 231 | 63 |
BC019208.1 | - | 1 - 2 | 177 | 63 | |
S80191.1 | - | 1 - 2 | 176 | 63 |