Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lad1
Entrez Gene ID
16763
Gene Name
ladinin
Gene Aliases
-
Chromosome Location
Chr.1: 135818598 - 135833341 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133664.3 NP_598425.2 9 - 10 1963 79
GenBank mRNA AK083526.1 - 9 - 10 1964 79
BC016257.1 - 9 - 10 1634 79
BC025580.1 - 9 - 10 1867 79
BC031131.1 - 9 - 10 1810 79

View Full Product Details