The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133694.2 | NP_598455.2 | 3 - 4 | 1003 | 84 |
GenBank mRNA | AK145009.1 | - | 3 - 4 | 963 | 84 |
AK172339.1 | - | 4 - 5 | 900 | 84 | |
AW124512.1 | - | 1 - 2 | 84 | ||
AW825726.1 | - | 1 - 2 | 409 | 84 | |
BC016499.1 | - | 3 - 4 | 978 | 84 |