The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133726.2 | NP_598487.1 | 2 - 3 | 263 | 75 |
GenBank mRNA | AK013981.1 | - | 2 - 3 | 250 | 75 |
AK146528.1 | - | 2 - 3 | 259 | 75 | |
AK147169.1 | - | - | 286 | 75 | |
AK152270.1 | - | 2 - 3 | 259 | 75 | |
AK154658.1 | - | - | 252 | 75 | |
AK159149.1 | - | - | 243 | 75 | |
BC003843.1 | - | 2 - 3 | 263 | 75 | |
CT010318.1 | - | 2 - 3 | 163 | 75 |