The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199009.1 | NP_001185938.1 | 13 - 14 | 1659 | 73 |
NM_133734.3 | NP_598495.1 | 13 - 14 | 1564 | 73 | |
NR_037572.1 | 13 - 14 | 1676 | 73 | ||
GenBank mRNA | AK033540.1 | - | 13 - 14 | 1562 | 73 |
AK150183.1 | - | 13 - 14 | 1651 | 73 | |
AK152395.1 | - | 12 - 13 | 1617 | 73 | |
AK158784.1 | - | 13 - 14 | 1644 | 73 | |
AK170657.1 | - | 13 - 14 | 1562 | 73 | |
BC008545.1 | - | 12 - 13 | 1457 | 73 | |
BC021305.1 | - | 10 - 11 | 3473 | 73 | |
BC037001.1 | - | 12 - 13 | 1588 | 73 |